Learn More
Sigma Aldrich Fine Chemicals Biosciences Hartmans fixative histolog1G
Catalog No. 502904030
Encompass
Hartman fixative which is also known as Davidson fixative contains acetic acid alcohol and formalin. It is widely used for the preservation of eye bone marrow breast testis and other tissues for histologic examination.
Catalog No. 50-290-4030 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H02901GAL
Hartman fixative which is also known as Davidson fixative contains acetic acid alcohol and formalin. It is widely used for the preservation of eye bone marrow breast testis and other tissues for histologic examination.