Learn More
3M Harvest RC, BC16000, 16 in. 7-Cell
Catalog No. 02557956 Shop All Thermo Scientific Products
This item is not returnable. View return policy
The brand will transition from 3M to Thermo Scientific
Catalog No. 02-557-956 Supplier 3M Supplier No. EMP770HRC2FA
Add to Cart
Add to Cart
This item is not returnable. View return policy
The brand will transition from 3M to Thermo Scientific
The Harvest RC Chromatographic Clarifier is a single‐use, single‐stage option for efficient harvesting and clarification of modern cell cultures. It streamlines processing with fewer steps, higher yield, and reduced production time and cost.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA (<500 ppb) from the harvested cell culture fluid.
  • Optimized for PCV high density CHO cell culture (5 to 8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Specifications
Disposable Single-use
Filter Type Filter Capsule
Material PES
Pore Size 0.2 μm
Sterility Non-sterile
Type Chromatographic Clarifier