Learn More
Description
- Single-stage, single-use chromatographic clarifier for harvesting and clarification of cell cultures.
- Uses synthetic fibrous anion exchange (AEX) chromatography media with a 0.2 μm polyether sulfone (PES) membrane.
- Removes cells, cell debris and soluble impurities, including DNA to <500 ppb.
- Designed for low- to high-density cell cultures, including >40 million cells/mL.
- Optimized for high-density CHO cell culture with 5 to 8% packed cell volume (PCV).
- Typical product recovery of ≥95% for capsule formats.
- Can replace primary, secondary and guard membrane clarification stages.
- Synthetic chromatographic media has chemically defined extractables.
- Supports predictable scaling from discovery through manufacturing.
- Uses less buffer and water compared with depth filtration.
- Available in capsule formats for laboratory through manufacturing-scale workflows.
Specifications
Specifications
| Filter Type | Bottle Top Filter |
| Pore Size | 0.2 μm |
| Model | BT500 |
| Quantity | 6/Cs. |
| Type | Chromatographic Clarifier |
| Volume (Metric) Working | 500 mL |
| Height (Metric) | 16.1 cm |
| Diameter (Metric) | 11.1 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.