Learn More
Chemglass Life Sciences Harvest Tube, M10, for 5L Bio Reactors
SDP
Catalog No. NC1673308
Encompass
Harvest Tube, M10, for 5L Bio Reactors. Harvest tubes mount into the outer M10 or M27 thread on the standard head plate. The design has the tapered tip within 2mm of the bottom of the bio reaction vessel to allow complete harvesting of all cells. The 15 Liter size has an M27 thread with 1/2” tubing for quicker harvest. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. NC1673308 Supplier Chemglass Life Sciences Supplier No. CLS138212
Harvest Tube, M10, for 5L Bio Reactors. Harvest tubes mount into the outer M10 or M27 thread on the standard head plate. The design has the tapered tip within 2mm of the bottom of the bio reaction vessel to allow complete harvesting of all cells. The 15 Liter size has an M27 thread with 1/2” tubing for quicker harvest. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.