Learn More
Chemglass Life Sciences Chemglass Harvest Tube, M10 for 3 L Bioreactors
SDP
Catalog No. 502733323
Encompass
Harvest tube designed to mount into the outer M10 headplate port of 3 L bioreactors. The tapered tip sits within 2 mm of the vessel bottom to enable near-complete harvesting of culture fluid and cells. Constructed of 316L stainless steel with an electropolished finish for corrosion resistance and cleanability.
  • Fits M10 headplate ports on 3 L bioreactors.
  • Tapered tip within 2 mm of vessel bottom for complete harvesting.
  • 316L stainless steel components.
  • Electropolished surface finish of 20 Ra or better.
  • Supplies like-for-like replacement for standard bioreactor harvest ports.
Catalog No. 50-273-3323 Supplier Chemglass Life Sciences Supplier No. CLS138032
Harvest tube designed to mount into the outer M10 headplate port of 3 L bioreactors. The tapered tip sits within 2 mm of the vessel bottom to enable near-complete harvesting of culture fluid and cells. Constructed of 316L stainless steel with an electropolished finish for corrosion resistance and cleanability.
  • Fits M10 headplate ports on 3 L bioreactors.
  • Tapered tip within 2 mm of vessel bottom for complete harvesting.
  • 316L stainless steel components.
  • Electropolished surface finish of 20 Ra or better.
  • Supplies like-for-like replacement for standard bioreactor harvest ports.