Learn More
Chemglass Life Sciences Chemglass Harvest Tube, M10, for 7 L Bioreactors (316L stainless steel)
SDP
Catalog No. 502733311
Encompass
Stainless-steel harvest tube with M10 thread designed for 7 L bioreactors. The tapered tip sits within 2 mm of the vessel bottom to enable complete harvesting of cells and media, and the electropolished finish aids cleanability and corrosion resistance.
  • 316L stainless steel construction.
  • M10 threaded connection.
  • Tapered tip positioned within 2 mm of the vessel bottom for complete harvest.
  • Electropolished surface finish (20 Ra) for improved cleanability.
  • Designed for 7 L bioreactors.
  • Sold each.
Catalog No. 50-273-3311 Supplier Chemglass Life Sciences Supplier No. CLS138313
Stainless-steel harvest tube with M10 thread designed for 7 L bioreactors. The tapered tip sits within 2 mm of the vessel bottom to enable complete harvesting of cells and media, and the electropolished finish aids cleanability and corrosion resistance.
  • 316L stainless steel construction.
  • M10 threaded connection.
  • Tapered tip positioned within 2 mm of the vessel bottom for complete harvest.
  • Electropolished surface finish (20 Ra) for improved cleanability.
  • Designed for 7 L bioreactors.
  • Sold each.