Learn More
AdipoGen Harzianopyridone, 137813-88-8, MFCD23160296, 1 mg
SDP
Supplier:  AdipoGen AGCN20149M001
Encompass_Preferred
MF: C14H19NO5, MW: 281.3, ≥95%, Appearance: White solid. Antibiotic. Antifungal and antibacterial compound. Specific mitochondrial complex II (succinate ubiquinone oxidoreductase; succinate dehydrogenase) inhibitor. Herbicidal activity. Anthelmintic compound. Inhibits NADH-fumarate reductase activity of adult Ascaris suum mitochondria.
Catalog No. 50-117-5983