Learn More
AdipoGen Harzianopyridone (250 µg)
SDP
Supplier:  AdipoGen AGCN20149C250
Encompass_Preferred
Chemical. CAS: 126637-69-2. Formula: C14H19NO5. MW: 281.3. Isolated from Trichoderma harzianum FKI-1509. Antibiotic. Antifungal and antibacterial compound. Specific mitochondrial complex II (succinate ubiquinone oxidoreductase; succinate dehydrogenase) inhibitor. Herbicidal activity. Anthelmintic compound. Inhibits NADH-fumarate reductase activity of adult Ascaris suum mitochondria.
Catalog No. 50-205-3530