Learn More
AdipoGen Harzianum A (1 mg)
SDP
Supplier:  AdipoGen AGCN20117M001
Encompass_Preferred
Chemical. CAS: 156250-74-7. Formula: C23H28O6. MW: 400.4. Isolated from Trichoderma sp. Antifungal. Anticancer compound. Cytotoxic to human cancer cell lines.
Catalog No. 50-205-3483