Learn More
Cayman Chemical Harzianum A, Purity: ≥95%, CAS: 156250-74-7, 10 mg
SDP
Catalog No. 502941526
Encompass_Preferred
A trichothecene fungal metabolite; is cytotoxic to HeLa, MCF-7, and HT-1080 cells (IC50s = 5.07, 10.13, and 0.65 μg/ml, respectively); exhibits antifungal activity against C. albicans and S. cerevisiae at 100 μg/ml
Catalog No. 50-294-1526 Supplier Cayman Chemical Supplier No. 2774010 MG
A trichothecene fungal metabolite; is cytotoxic to HeLa, MCF-7, and HT-1080 cells (IC50s = 5.07, 10.13, and 0.65 μg/ml, respectively); exhibits antifungal activity against C. albicans and S. cerevisiae at 100 μg/ml