Learn More
Aviva Systems Biology Corp HAS1 Antibody - N-terminal region (ARP63382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAS1. It was validated on Western Blot. . Alternate names: HASApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-953