Learn More
Eurofins Discoverx Haspin, active, 10 µg. (14-744)
Supplier:  Eurofins Discoverx 14744
Encompass
N-terminal 6xHis-tagged recombinant, human Haspin amino acids 471-end. Histone and Histone-Modifying Enzyme. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9275