Learn More
Analytical Sales & Services Hastelloy C22, Plug, ZDV, 1/8''
Catalog No. 5000806812
Encompass
Hastelloy C22, Plug, ZDV, 1/8''\n\nCan be used as a plug for one end of a union, or for any other tubing ports that are standard for 1/8" OD tubing.
Catalog No. 50-008-06812 Supplier Analytical Sales & Services Supplier No. HCP018
Add to Cart
Add to Cart
Hastelloy C22, Plug, ZDV, 1/8''\n\nCan be used as a plug for one end of a union, or for any other tubing ports that are standard for 1/8" OD tubing.