Learn More
Analytical Sales & Services Hastelloy C22 Reducing Union, 1/4" to 1/8"
Catalog No. 5000807235
Encompass
 Hastelloy C22 Reducing Union (HC22 Body and Ferrules, 316SS Nuts)\n• 1/4" to 1/8"\n• 0.080 thru-hole\n\nQty/Pkg: Each
Catalog No. 50-008-07235 Supplier Analytical Sales & Services Supplier No. HCRU1418L
 Hastelloy C22 Reducing Union (HC22 Body and Ferrules, 316SS Nuts)\n• 1/4" to 1/8"\n• 0.080 thru-hole\n\nQty/Pkg: Each