Learn More
Analytical Sales & Services Hastelloy C22, ZDV, Tee, 1/32", 0.010" Thru-hole
Catalog No. 5000805564
Encompass
Hastelloy C22, ZDV, Tee, 1/32", 0.010" Thru-hole\n\nQty/Pkg: Each
Catalog No. 50-008-05564 Supplier Analytical Sales & Services Supplier No. HCT132
Hastelloy C22, ZDV, Tee, 1/32", 0.010" Thru-hole\n\nQty/Pkg: Each