Learn More
Sigma Aldrich Hastelloy™ C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
Catalog No. 503092112 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-2112 1 Ea.
1 options
Catalog No. 50-309-2112 Supplier Sigma Aldrich Supplier No. GF488622991EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Hastelloy™ C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
Specifications
Quantity 1 Ea.
Synonym NI140271