Learn More
Sigma Aldrich Hastelloy™ C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
Catalog No. 503093132 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 Ea.
Catalog No. Quantity
50-309-3132 10 Ea.
1 options
Catalog No. 50-309-3132 Supplier Sigma Aldrich Supplier No. GF3816782510EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Hastelloy™ C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
Specifications
Quantity 10 Ea.
Synonym NI140270