Learn More
Fisherbrand™ Hastelloy External Probe
Catalog No. CIC0001467 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Isotemp Hotplates and Hotplate Stirrers
Catalog No. For Use With (Equipment)
CIC0001467 Isotemp Hotplates and Hotplate Stirrers
1 options
Catalog No. CIC0001467
Add to Cart
Add to Cart
For Use With: Isotemp Hotplates and Hotplate Stirrers
Hastelloy PT1000 Temperature probe for use with Isotemp and Isotemp Advanced hotplates and hotplate stirrers
  • Hastelloy PT1000 Temperature probe
  • Temp. Range: 0 to 400°C
  • Chemical and corrosion resistant
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Isotemp Hotplates and Hotplate Stirrers
Temperature (Metric) 0°C to 400°C
Type Probe