Learn More
Anton Paar HASTELLOY C-276, LUER ADAPTER, FOR PELTIER, 100 mm for MCP 100
Catalog No. NC1818310
Encompass_Preferred
100 mm hastelloy sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 100.Filling volume approximately 0.7 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306
Catalog No. NC1818310 Supplier Anton Paar Supplier No. 143947
100 mm hastelloy sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 100.Filling volume approximately 0.7 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306