Learn More
Selleck Chemical LLC Histone Acetyltransferase Inhibitor II S1161-5mg
SDP
Catalog No. 503111375
Encompass_Preferred
Histone Acetyltransferase Inhibitor II (HAT Inhibitor II compound 2c) is a potent selective and cell-permeable p300 histone acetyltransferase (HAT) inhibitor with IC50 of 5 M Histone Acetyltransferase Inhibitor II shows anti-acetylase activity in mammalian cells
Catalog No. 50-311-1375 Supplier Selleck Chemical LLC Supplier No. S11615MG
Add to Cart
Add to Cart
Histone Acetyltransferase Inhibitor II (HAT Inhibitor II compound 2c) is a potent selective and cell-permeable p300 histone acetyltransferase (HAT) inhibitor with IC50 of 5 M Histone Acetyltransferase Inhibitor II shows anti-acetylase activity in mammalian cells