Learn More
Cayman Chemical HAT Inhibitor II, Purity: ≥98%, CAS: 932749-62-7, 5 mg
SDP
Catalog No. 502982425
Encompass_Preferred
A cell-permeable, selective inhibitor of the HAT p300 (IC50 = 5 µM)
Catalog No. 50-298-2425 Supplier Cayman Chemical Supplier No. 198355 MG
A cell-permeable, selective inhibitor of the HAT p300 (IC50 = 5 µM)