Learn More
Apexbio Technology LLC HATU, 25g. Cas: 148893-10-1 MFCD: MFCD00274639. used in peptide coupling chemistry
SDP
Catalog No. 501904966
Encompass_Preferred
HATU(1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used inpeptide couplingchemistry to generate an active ester from a carboxylic acid. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4966 Supplier Apexbio Technology LLC Supplier No. A702225000
HATU(1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used inpeptide couplingchemistry to generate an active ester from a carboxylic acid. Other sizes are available. Please inqury us for quote.