Learn More
Aviva Systems Biology Corp HAUS8 Antibody - N-terminal region (ARP59892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAUS8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HAUS8. Alternate names: HICE1, MGC20533, NY-SAR-48, DGT4ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-286-869