Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hausser Scientific™ Nageotte Bright-Line™ Hemacytometer
GSA_VA

Catalog No. 0267152
Change view
Click to view available options
Depth (English):
0.019 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
02-671-52 0.019 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-671-52 Supplier Hausser Scientific™ Supplier No. 4000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manually count low leukocyte concentrations below the capabilities of a standard hemacytometer. Hausser Nageotte Bright-Line Hemacytometer can accurately measure down to 0.1 cell/mL.

  • Allows manual counting of low leukocyte concentrations below the capabilities of standard hemacytometers
  • Contains two gridded counting areas each containing 40 rectangular areas
  • Total volume is 50mL for a total volue of 100mL per Nageotte
  • Can accurately measure down to 100 cell/mL
  • 0.5mm cell depth
  • H-shaped moat that separates the two gridded areas 
  • Two reusable cover slips (30 x 33 x 0.5mm) and 1/2 oz (approx. 35 pcs.) of disposable cover slips (30 x 33 x 0.15mm)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Hemacytometer
Grid Type Nageotte
Capacity (Metric) 100μL
Depth (English) 0.019 in.
Depth (Metric) 0.05cm
For Use With (Application) Counting leukocyte concentrations
Includes Two reusable cover slips (30 x 33 x 0.5mm) and 1/2 oz (approx. 35 pcs.) of disposable cover slips (30 x 33 x 0.15mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.