Learn More
Hausser Scientific™ Hemocytometer
SCI_ED
Catalog No. S17040
Change view
Click to view available options
For Use With (Application):
Tool for Blood tests
Catalog No. For Use With (Application)
S17040 Tool for Blood tests
1 options
This item is not returnable. View return policy
Catalog No. S17040 Supplier Hausser Scientific™ Supplier No. A01483 S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
  • Bright Line Chamber with double Neubauer rulings
  • Two cover glass
  • One red pipette
  • One white pipette
  • One pipette filler
  • Clear case
Get quick, accurate readings of red and white blood cells.
  • Easy-to-read, brightline Neubauer counting chamber makes device an important tool for blood tests
  • Made from one piece of optical glass, its double ruling allows check counts to be made for accuracy
Specifications
Product Type Hemacytometer
Grid Type Neubauer Bright Line
Quantity Each
For Use With (Application) Tool for Blood tests
Includes Two cover Glass, one red pipette, one white pipette, one pipette filler