Learn More
Creative Biomart Recombinant HAV P3C Antigen
Catalog No. 502084462
Encompass_Preferred
The E. coli derived 40 kDa recombinant protein contains the P3C immunodominant regions, amino acids 1643-1743.
Catalog No. 50-208-4462 Supplier Creative Biomart Supplier No. HAV084
The E. coli derived 40 kDa recombinant protein contains the P3C immunodominant regions, amino acids 1643-1743.