Learn More
Aviva Systems Biology Corp HAVCR2 Antibody - C-terminal region (ARP59159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAVCR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HAVCR2. Alternate names: FLJ14428, KIM-3, TIM3, TIMD3, Tim-3, TIMD-3, HAVcr-2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-286-599