Learn More
Aviva Systems Biology Corp HAVCR2 Antibody - N-terminal region (ARP45550_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAVCR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HAVCR2. Alternate names: FLJ14428, KIM-3, TIM3, TIMD3, Tim-3, TIMD-3, HAVcr-2ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-59050