Learn More
Haws™ Anti-Freeze Drench Shower
SDP
Catalog No. 18888217 Shop All Haws Corporation Products
Change view
Click to view available options
Mounting:
Wall
Catalog No. Mounting
18-888-217 Wall
1 options
Catalog No. 18-888-217 Supplier Haws™ Supplier No. 8111FP
  • $1,624.00
    $1,270.65 / Each of 1
    Save $353.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Anti-Freeze Drench Shower features an AXION™ MSR showerhead.
  • Excellent addition to multiple potentially unsafe environments where the body may be exposed to corrosive materials.
  • AXION MSR ABS plastic drench showerhead with flow control delivers dispersed flow for uniform drenching.
  • Requires minimal maintenance and is equipped with test card to record weekly testing.
  • Reducing installation time, valve assembly is pre-built
Warranty and Services
1 year from ship date (Limited)
Specifications
Certifications/Compliance ANSI Z358.1
Flow Rate 20 gpm
Inlet Connection (English) 1 in.
Inlet Connection (Metric) 2.54
Material Plastic
Mounting Wall
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov