Learn More
MSC Haws AXION eyePOD Faucet Eyewash, Inverted Flow Design, 1 EA
Catalog No. 5000674834
Encompass
This faucet-mounted eyewash features a revolutionary inverted flow design that provides a medically superior response by sweeping contaminants away from the vulnerable nasal cavity, consistent with EMT, emergency room, and doctor office protocols.
  • Eyewash streams provide zero vertical velocity stream engineering with even laminar flow for enhanced comfort, stability, and effectiveness.
  • Thermostatic shutoff protects the user from water over 100° F (37.8° C) by shutting off the eyewash.
  • Supplied with a standard 55/64-27 female connection and multiple commonly used faucet adapters.
  • Constructed of stainless steel with a polished finish for durability and aesthetics.
  • Dimensions of 3.5 in. (8.9 cm) diameter and 2 in. (5 cm) height.
  • Certified to meet the ANSI/ISEA Z358.1 Standard for Emergency Eyewash and Shower Equipment.
Catalog No. 50-006-74834 Supplier MSC Supplier No. 51889160
This faucet-mounted eyewash features a revolutionary inverted flow design that provides a medically superior response by sweeping contaminants away from the vulnerable nasal cavity, consistent with EMT, emergency room, and doctor office protocols.
  • Eyewash streams provide zero vertical velocity stream engineering with even laminar flow for enhanced comfort, stability, and effectiveness.
  • Thermostatic shutoff protects the user from water over 100° F (37.8° C) by shutting off the eyewash.
  • Supplied with a standard 55/64-27 female connection and multiple commonly used faucet adapters.
  • Constructed of stainless steel with a polished finish for durability and aesthetics.
  • Dimensions of 3.5 in. (8.9 cm) diameter and 2 in. (5 cm) height.
  • Certified to meet the ANSI/ISEA Z358.1 Standard for Emergency Eyewash and Shower Equipment.