Learn More
Haws™ AXION MSR™ Swing-Action Eye/Face Washes
SDP
Catalog No. 19033729 Shop All Haws Corporation Products
Change view
Click to view available options
Mounting:
Rear
Catalog No. Mounting
19-033-729 Rear
1 options
Catalog No. 19-033-729 Supplier Haws™ Supplier No. 7610
Add to Cart
Add to Cart
Includes
Universal language emergency sign and waterproof test tag
Polished chrome-plated lab units with swing-down or swing-away action
  • Ideal for areas where side counter space is limited, Model 7610 can be deck or wall mounted with easy change-over instructions
  • 7610 activated by swinging unit down over the sink, the rear mount design allows the sink to remain open to other uses
  • Low-profile Model 7611 is mounted to either the right or left side of sink, activated by swinging sideways over the sink
  • Eye/face wash streams provide zero vertical velocity stream engineering with even laminar flow for enhanced comfort, stability and effectiveness
  • A single motion activates unique stay-open ball valve to release spray
  • Supply, .5 in. O.D. slip joint
Certifications
CSA certified to meet ANSI™ Z358.1
Specifications
Certifications/Compliance ANSI Z358.1
For Use With (Application) Sweeping Contaminants away from the Vulnerable Nasal Cavity
Includes ABS Plastic Eyewash Head, Dust Covers, Universal Language, Emergency Sign, Waterproof Test Tags, Extended-length Pull Down Arms, Chrome-plated
Mounting Rear
Product Type Swing Down Eye and Face Wash
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov