Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Sterile Bacteriostatic Water Preservative
SDP

Catalog No. 1798710 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Portable Eyewash Station
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-987-10 Portable Eyewash Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-10 Supplier Haws™ Supplier No. 9082
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Model 9082, 'sterile' bacteriostatic preservative for use in portable eyewash stations. One 5 oz. (148 ml) bottle will preserve 5 to 20 gallons (18.9 to 75.7 L) of potable water for up to three months. Four bottles per carton.

  • Model 9082, 'sterile' bacteriostatic preservative for use in portable eyewash stations.
  • One 8 oz. (237 ml) bottle will preserve 5 to 20 gallons (18.9 to 75.7 L) of potable water for up to 120 days.
  • This sterile solution is in compliance with FDA regulations.
  • Temperature storage requirements; store at 36-104°F, and use at 59-86°F.
  • Shelf life of 36 months
  • Four bottles per carton.

Specifications

Capacity (English) 8 oz.
Capacity (Metric) 237 mL
For Use With (Equipment) Portable Eyewash Station
Product Type Bacteriostatic Water Preservative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.