Learn More
Haws™ Ball Valve
SDP
Catalog No. 18888226 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
18-888-226 Stainless Steel
1 options
Catalog No. 18-888-226 Supplier Haws™ Supplier No. SP225LF
  • $338.00
    $268.65 / Each of 1
    Save $69.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Ball Valve is a chrome-plated, brass, stay-open ball valve, equipped with a stainless steel ball and stem
  • Push flag not included
  • 1/2-inch inlet x 1/4-inch NPT outlet
Warranty and Services
1 year from ship date (Limited)
Specifications
Material Stainless Steel
Product Type Ball Valve
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov