Learn More
Haws™ Bubbler Valves
SDP
Catalog No. 19133777 Shop All Haws Corporation Products
Change view
Click to view available options
Model:
Model 5701SL
Model 5703M
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-133-777 Model 5703M
19-133-776 Model 5701SL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-133-777 Supplier Haws™ Supplier No. 5703M
  • $742.00
    $589.65 / Each of 1
    Save $152.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Model 5701SL & 5703M, polished chrome-plated forged brass, shielded, anti-squirt bubbler head with vandal-resistant mounting slot.
Model 5703M is a polished chrome-plated brass bubbler head with shielded sanitary nozzle design, mounting hardware, and a 3/8 in. compression inlet with 100% lead-free waterways.
Specifications
Certifications/Compliance Industry Standards
For Use With (Application) Provide an Anti-squirt Drinking Stream
Model Model 5703M
Product Type Bubbler Valve