Learn More
Haws™ Chrome-Plated Brass Ball Valve With Flag, Self-Draining
SDP
Catalog No. 18888230 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Chrome-plated Brass
Catalog No. Material
18-888-230 Chrome-plated Brass
1 options
Catalog No. 18-888-230 Supplier Haws™ Supplier No. SP229.158
  • $1,105.00
    $877.65 / Each of 1
    Save $227.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Chrome-Plated Brass Ball Valve With Flag, Self-Draining is equipped with a stainless steel ball and stem.
Self-draining feature allows valve and connecting lines to eyewash to drain to prevent freezing.
Warranty and Services
1 year from ship date (Limited)
Specifications
Includes Flag
Material Chrome-plated Brass
Product Type Ball Valve
Weight (English) 2 lb.
Weight (Metric) 0.91 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov