Learn More
Haws™ AXION™ MSR Emergency Drench Shower
SDP
Catalog No. 19038415 Shop All Haws Corporation Products
Change view
Click to view available options
Mounting:
Ceiling
Ceiling, Flush
Horizontal or Vertical
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
19-038-415 Ceiling, Flush
19-038-414 Ceiling
17-164B Horizontal or Vertical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-038-415 Supplier Haws™ Supplier No. 8164
  • $3,108.00
    $2,431.65 / Each of 1
    Save $676.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Outfitted with safety features like a self-regulating shower stream and universal emergency sign, the Haws AXION MSR Emergency Drench Showers are designed for safety.
Haws drench showers feature the AXION MSR hydrodynamic shower design that minimizes the strong physical pressure that drives water to the outer rim of normal shower heads, making the distribution equal across the entire footprint of the flow. Better use of the water flowing through the shower makes it more effective while providing a more comfortable environment by breaking the streams of water into smaller individual droplets.
Specifications
Certifications/Compliance ANSI Z358.1, CSA
Flow Rate 20 gpm
For Use With (Application) Quick Drenching or Cleansing of the Body
Includes AXION MSR™ hydrodynamic design polished Stainless Steel, Brass stay open ball valve, recessed Stainless Steel wall cabinet with pull down lever, universal sign
Mounting Ceiling, Flush
Product Type Drench Shower
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov