Learn More
Haws™ Eyewash Bend
SDP
Catalog No. 18888257 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
18-888-257 Stainless Steel
1 options
Catalog No. 18-888-257 Supplier Haws™ Supplier No. SP8
  • $126.00
    $93.65 / Each of 1
    Save $32.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Eyewash Bend is made from electropolished Type 304 stainless steel
Model SP8 electropolished Type 304 stainless steel eye/face wash bend. Used on eye and face wash head assemblies. 1/4 in. IPS.
Warranty and Services
1 year from ship date (Limited)
Specifications
Color Silver
Material Stainless Steel
Product Type Eyewash Bend
WARNING: Cancer - www.P65Warnings.ca.gov