Learn More
Haws™ Eyewash Gauge
SDP
Catalog No. 18888258 Shop All Haws Corporation Products
Change view
Click to view available options
Color:
Transparent
Catalog No. Color
18-888-258 Transparent
1 options
Catalog No. 18-888-258 Supplier Haws™ Supplier No. 9015
  • $41.50
    $35.65 / Each of 1
    Save $5.85
    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Eyewash Gauge allows easy testing of eyewashes per ANSI Standard Z358.1.
Model 9015 eyewash gauge allows for easy testing of eyewashes per ANSI Standard Z358.1. Directions are in black on the clear Acrylic. Model 9015 also serves as a 6 in. (15 cm) ruler.
Warranty and Services
1 year from ship date (Limited)
Specifications
Color Transparent
Product Type Eyewash Gauge