Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Eyewash Head
SDP

Catalog No. 18888259 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
7500, 7500EB, 7501, 7501T
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-259 7500, 7500EB, 7501, 7501T
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-259 Supplier Haws™ Supplier No. SP19
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Eyewash Head features a tripod anchor, 8-32 stainless steel socket head cap screw and rubber gasket.

Model SP19 is a ABS plastic eyewash head with tripod anchor, 8-32 stainless steel socket head cap screw and rubber gasket.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Silver
For Use With (Application) Eyewash
For Use With (Equipment) 7500, 7500EB, 7501, 7501T
Material Stainless Steel, Plastic
Product Type Eyewash Head
Weight (English) 1 lb.
Weight (Metric) 0.453592 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.