Learn More
Haws™ Freeze Valve
SDP
Catalog No. 18888269 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Emergency Showers
Emergeny Shower and Eyewash
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-269 Emergeny Shower and Eyewash
18-888-270 Emergency Showers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-888-269 Supplier Haws™ Supplier No. SP158.8
Add to Cart
Add to Cart
Haws™ Freeze Valve is a retro-fit freeze protection valve kit
Model SP158.8, retro-fit freeze valve kit to replace the stainless steel freeze valve on Haws model 8300.158. This kit is supplied with the necessary hardware and instructions.
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) Emergeny Shower and Eyewash
Material Brass
Product Type Freeze Valve
Weight (English) 3 lb.
Weight (Metric) 1.36 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov