Learn More
Haws™ Freeze Valve
SDP
Catalog No. 18888270 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Emergency Showers
Emergeny Shower and Eyewash
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-270 Emergency Showers
18-888-269 Emergeny Shower and Eyewash
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-888-270 Supplier Haws™ Supplier No. SP158CTFP
  • $1,534.00
    $1,218.65 / Each of 1
    Save $315.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Freeze Valve is a retro-fit freeze protection valve kit
Model SP158CTFP, retro-fit freeze valve kit to replace the stainless steel valve on Haws model 8317CTFP. This kit is supplied with the necessary hardware, insulation and replacement instructions.
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) Emergency Showers
Material Brass
Product Type Freeze Valve
Weight (English) 3 lb.
Weight (Metric) 1.36 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov