Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Green Area Light
SDP

Catalog No. 18888271 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8317CTFP, Emergency Shower
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-271 8317CTFP, Emergency Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-271 Supplier Haws™ Supplier No. 8317LT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Green Area Light is for continuous illumination.

Model 8317LT is a vapor tight and gasketed green light for continuous illumination of the 8317CTFP series showers.

  • CSA electrical certified, TYPE 4. For Class 1, Div. 2, Group B, C & D.
  • Temperature Code: T2C; only when used with 8317CTFP.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Green
For Use With (Equipment) 8317CTFP, Emergency Shower
Product Type Watertight Area Light
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.