Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Horizontal Flag
SDP

Catalog No. 18888275 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Horizontally Mounted Eyewash Ball Valve
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-275 Horizontally Mounted Eyewash Ball Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-275 Supplier Haws™ Supplier No. SP216
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Horizontal Flag is a stainless steel push flag with graphics for use on horizontally mounted eyewash ball valves.

Model SP216 is a push flag for eyewash valves.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Silver
For Use With (Equipment) Horizontally Mounted Eyewash Ball Valve
Material Stainless Steel
Product Type Horizontal Flag
Weight (English) 1 lb.
Weight (Metric) 0.453592 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.