Learn More
Haws™ Hose Units
SDP
Catalog No. 19811866 Shop All Haws Corporation Products
Change view
Click to view available options
Mounting:
Wall
Catalog No. Mounting
19-811-866 Wall
1 options
Catalog No. 19-811-866 Supplier Haws™ Supplier No. 8901B
  • $972.00
    $759.65 / Each of 1
    Save $212.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Includes
Wall mounting bracket.
Assists in irrigation process
  • Dust cover allows for spray head to stay protected from debris when not in use
  • Requires minimal maintenance and is equipped with test card to record weekly testing
  • 8 ft. (243.8cm) swivel hose provides extended coverage area to further optimiz irrigation process
  • Drench head can be easily activated with integral squeeze lever
  • Conveniently mounted using wall clips
Compliance
Certified by CSA to meet the ANSI Z358.1 Standard for Emergency Eyewash and Shower Equipment
Specifications
Certifications/Compliance ANSI Z358.1, CSA
For Use With (Application) Maximum Coverage area for the Eye, Face, or Body
Includes Green ABS Plastic head, Chrome Plated Brass stay open squeeze lever valve, Green Rubber hose, wall mounting bracket, universal sign
Length (English) 8 ft.
Length (Metric) 2.43 m
Mounting Wall
Product Type Hose Unit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov