Learn More
Haws™ In-Line Strainer
SDP
Catalog No. 18888279 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Chrome-plated Brass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-888-279 Stainless Steel
18-888-278 Chrome-plated Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-888-279 Supplier Haws™ Supplier No. SP509SS
  • $541.00
    $405.65 / Each of 1
    Save $135.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ In-Line Strainer is a 50x50 mesh strainer that filters debris from reaching the eye or eye/face wash
Model SP509SS is a Type 304 stainless steel in-line 50 x 50 mesh water strainer that prevents debris from reaching the eye/face wash. Easily serviceable.
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) 7777, 7778B, Eye and Face Wash
Material Stainless Steel
Product Type In-line Strainer
Weight (English) 1 lb.
Weight (Metric) 0.453592 kg
WARNING: Cancer - www.P65Warnings.ca.gov