Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Mounting Bracket
SDP

Catalog No. 18888284 Shop All Haws Corporation Products
Change view
Click to view available options
Mounting:
Wall
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
18-888-284 Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-284 Supplier Haws™ Supplier No. SP80
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Mounting Bracket features a chromate protective finish.

Model SP80 is a cast aluminum mounting bracket used with wall mounted eye and eye/face wash models.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Cromate Finish
For Use With (Equipment) 7260B-7270B, 7260BT-7270BT, 7360B-7460B, 7360BT-7460BT, 7433FP
Material Cast Aluminum
Mounting Wall
Weight (English) 2 lb.
Weight (Metric) 0.907185 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.