Learn More
Haws™ Omni-Flo™ Combination Unit
SDP
Catalog No. 17988384 Shop All Haws Corporation Products
Change view
Click to view available options
Eye/Facewash and Shower Unit
Catalog No.
17-988-384 Eye/Facewash and Shower Unit
1 options
Catalog No. 17-988-384 Supplier Haws™ Supplier No. 8200
Includes
Self-adhesive 21 in. long (53.3cm) stripe in Safety Green and bright yellow.
Combination eye/facewash and independently operated drench shower
Activation
  • Stainless-steel pull rod activates shower
  • Eyewash can be activated with foot treadle, for hands-free operation, or with large push flag
Shower Head
  • ABS plastic shower head is equipped with 20 gpm (75.7L/min.) flow control to ensure steady flow when flowing pressure is between 30 and 90psi
Eye/facewash Head and Bowl
  • Wraparound sprayhead design delivers a delicate, soothing spray of water over entire face
  • Eye/facewash control has 6.25 gpm (23.7L/min.) flow control; controls water flow rate when flowing pressure is between 30 and 90psi
  • Bright yellow ABS dust cover protects sprayhead until activated by water flow
  • Durable green ABS plastic receptor is resistant to damage from alkalis, salt solution, oils, most acids, sun exposure and color change
Pipe and Fittings
  • 1 1/4 in. (3.18cm) Schedule 40 hot-dipped galvanized steel
  • Features a gray, powder-coated 9 in. dia. (22.9cm) cast-iron floor flange
  • Supply and waste, 1 1/4 in. IPS
Certifications
CSA certified to ANSI™ Z358.1-2004.