Learn More
Haws™ Polished Chrome-Plated Brass Ball Valve With Flag
SDP
Catalog No. 18888227 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Polished Chrome-plated Brass
Catalog No. Material
18-888-227 Polished Chrome-plated Brass
1 options
Catalog No. 18-888-227 Supplier Haws™ Supplier No. SP226PCP
  • $818.00
    $650.65 / Each of 1
    Save $167.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Polished Chrome-Plated Brass Ball Valve With Flag is equipped with a stainless steel ball and stem
  • Designed for vertical installation
  • Full flow occurs with push on stainless steel flag
  • 0.5in inlet x 3/8in NPT outlet
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) Sink Mounted Eye and Face Wash
Includes Flag
Material Polished Chrome-plated Brass
Product Type Ball Valve
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov