Learn More
Haws™ Portable Eyewash Cart
SDP
Catalog No. 19033743 Shop All Haws Corporation Products
Change view
Click to view available options
Includes:
Ratchet Tie Down Strap
Catalog No. Includes
19-033-743 Ratchet Tie Down Strap
1 options
Catalog No. 19-033-743 Supplier Haws™ Supplier No. 9008
  • $1,166.00
    $872.65 / Each of 1
    Save $293.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Model 9008 cart designed to transport all stainless steel portable eyewashes.
Model 9008 cart designed to transport all stainless steel portable eyewashes. Unit is constructed of heavy-duty 1 in. (2.5 cm) tubular frame with green powder-coated finish, features rubber wheels, and is equipped with a ratchet tie down strap to securely hold the tank to the cart.
Specifications
Includes Ratchet Tie Down Strap
Material Stainless Steel
Product Type Portable Eyewash Cart
WARNING: Cancer - www.P65Warnings.ca.gov