Learn More
MSC Haws Portable Eyewash Station, Polyethylene, 16 gal, 0.4 GPM
Catalog No. 5000796884
Encompass
The Haws 7500 portable eyewash station is a gravity-fed unit designed for emergency eye rinsing. Constructed from durable polyethylene, it provides a 0.4 GPM flow rate for 15 minutes, meeting ANSI Z358.1-2014 standards.
  • Durable polyethylene construction
  • Gravity-fed operation
  • Capacity of 16 gal.
  • Provides a 0.4 GPM flow rate
  • Offers 15 minutes of continuous flow
  • Meets ANSI Z358.1-2014 standards
Catalog No. 50-007-96884 Supplier MSC Supplier No. 06116982
The Haws 7500 portable eyewash station is a gravity-fed unit designed for emergency eye rinsing. Constructed from durable polyethylene, it provides a 0.4 GPM flow rate for 15 minutes, meeting ANSI Z358.1-2014 standards.
  • Durable polyethylene construction
  • Gravity-fed operation
  • Capacity of 16 gal.
  • Provides a 0.4 GPM flow rate
  • Offers 15 minutes of continuous flow
  • Meets ANSI Z358.1-2014 standards